EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Regulation of the human endogenous retroviral Syncytin-1 and cell-cell fusion by the nuclear hormone receptors PPARγ/RXRα in placentogenesis.
PMID 22573555
Abstract
Cytotrophoblast (CT) cell fusion into a syncytiotrophoblast is obligatory for placentation and mediated by the human endogenous retrovirus (HERV)-W envelope gene Syncytin-1. Abnormal placentation is associated with preeclampsia (PE), HELLP and intrauterine growth restriction (IUGR). In placentogenesis, the MAP-kinase p38α regulates PPARγ/RXRα signaling and target genes, like leptin, resistin, ABCG2, and hCG. The aim of this study was to analyze PPARγ/RXRα signaling and target gene regulation using primary CT cultures, the trophoblastic cell line BeWo and placental tissues from patients with normal and abnormal placentation. CT from four different human control placentae and BeWo cells demonstrated that Syncytin-1, other signaling members and CT cell fusions were regulated with PPARγ/RXRα activators troglitazone and 9-cis retinoic acid, via protein kinase A and p38α inhibition. Significant discordant regulations between CTs and BeWo were found. Two PPARγ/RXRα-response-elements from upstream regulatory elements and the 5'LTR of HERV-W were confirmed with DNA-protein binding assays using nuclear extracts and recombinant PPARγ/RXRα proteins. These promoter elements were validated with luciferase assays in the presence of PPARγ/RXRα modulators. Furthermore, troglitazone or 9-cis retinoic acid treatment of siRNA-PPARγ and siRNA-RXRα transfected BeWo cells proved the requirement of these proteins for Syncytin-1 regulation. Thirty primary abnormal placentae from PE, HELLP and IUGR patients compared to 10 controls showed significant deregulation of leptin RNA and protein, p38α, phospho-p38α, PPARγ, ABCG2, INSL4 and Syncytin-1. Our study characterized PPARγ/RXRα signaling in human CT and cell fusions identifying Syncytin-1 as a new target gene. Based on these results, a disturbed PPARγ/RXRα pathway could contribute to pathological human pregnancies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4643
9-cis-Retinoic acid, ≥98% (HPLC)
C20H28O2