EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Identification of degradation products of thiram in water, soil and plants using LC-MS technique.
PMID 22575009
Abstract
In order to evaluate the deleterious effects of exposure to pesticides on a target population, a comprehensive study on their degradation in the various segments of ecosystem under varying environmental conditions is needed. In view of this, a study has been carried out on the metabolic pathways of thiram, a dithiocarbamate fungicide, in a variety of matrices namely water and soil under controlled conditions and plants in field conditions. The identification of degradation products was carried out in samples collected at various time points using LC-MS. The degradation products identified can be rationalized as originating by a variety of processes like hydrolysis, oxidation, N-dealkylation and cyclization. As a result of these processes the presence of some metabolites like dimethyl dithiocarbamate, bis(dimethyl carbamoyl) disulphide, bis(dimethyl dithiocarbamoyl) trisulphide and N-methyl-amino-dithiocarbamoyl sulphide was observed in all the cases. However, some different metabolites were observed with the change in the matrix or its characteristics such as cyclised products 2(N, N-dimethyl amino)thiazoline carboxylic acid and 2-thioxo-4-thiazolidine were observed only in plants. The investigations reflect that degradation initiates with hydrolysis, subsequently oxidation/dealkylation, followed by different types of reactions. The pathways seem to be complex and dependent on the matrices. Dimethyl dithiocarbamate and oxon metabolites, which are more toxic than parent compound, seem to persist for a longer time. Results indicate persistence vis-a-vis toxicity of pesticide and its metabolites and also provide a data bank of metabolites for forensic and epidemiological investigations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T24201
Tetramethylthiuram disulfide, 97%
C6H12N2S4
45689
Thiram, PESTANAL®, analytical standard
C6H12N2S4