EMAIL THIS PAGE TO A FRIEND
Nature protocols
Enzymatic incorporation of an azide-modified UTP analog into oligoribonucleotides for post-transcriptional chemical functionalization.
PMID 22576108
Abstract
This protocol describes the detailed experimental procedure for the synthesis of an azide-modified uridine triphosphate analog and its effective incorporation into an oligoribonucleotide by in vitro transcription reactions. Furthermore, procedures for labeling azide-modified oligoribonucleotides post-transcriptionally with biophysical probes by copper(I)-catalyzed alkyne-azide cycloaddition (CuAAC) and Staudinger reactions are also provided. This post-transcriptional chemical modification protocol is simple and modular, and it affords labeled oligonucleotides in reasonable amounts for biophysical assays. The procedure for enzymatic incorporation of the monophosphate of azide-modified UTP into an oligoribonucleotide transcript takes ∼2 d, and subsequent post-transcriptional chemical functionalization of the transcript takes about 2 d.