EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A new synthetic approach to benzoporphyrins and Kröhnke type porphyrin-2-ylpyridines.
PMID 22576564
Abstract
The reaction of 2-formyl-5,10,15,20-tetraphenylporphyrin with aryl methyl ketones and ammonium acetate, in the presence of La(OTf)(3), affords benzoporphyrins and 2-(2,6-diarylpyridin-4-yl)porphyrins. This methodology was used to prepare, for the first time, a 2-(2,2':6',2''-terpyridin-4'-yl)porphyrin. The structures of two 2-(2,6-diarylpyridin-4-yl)porphyrins were confirmed by single-crystal X-ray diffraction.
Related Materials