EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Ionization of shikonin derivatives using negative-ion electrospray mass spectrometry: [M-H]- versus [M + e]•-.
PMID 22576870
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7576
Shikonin, ≥98% (HPLC)
C16H16O5