EMAIL THIS PAGE TO A FRIEND
The Annals of thoracic surgery
Two consecutive open heart operations in a small child with heparin-induced thrombocytopenia type II using anticoagulation with heparin and tirofiban.
PMID 22579887
Abstract
We report the use of the platelet glycoprotein IIb/IIIa antagonist tirofiban for cardiac surgery in a small child with active heparin-induced thrombocytopenia type II, including description of the postoperative course.