EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Modular theranostic prodrug based on a FRET-activated self-immolative linker.
PMID 22580227
Abstract
New chemotherapeutic prodrugs that can report on the localization and activation of the drug upon internalization into select cells are being widely investigated for cancer treatment. Here, we introduce a new molecular design for a theranostic prodrug based on a self-immolative linker attached to a pair of FRET dyes that produces a fluorescent signal upon disassembly. The prodrug evaluated here was designed to release the chemotherapeutic drug camptothecin upon activation by the model enzyme penicillin-G-amidase. Similar patterns of the disassembly of the prodrug were observed in HPLC and fluorescence assays. The obtained results demonstrate that upon specific activation of the prodrug, the increase of the emitted fluorescent signal is linearly correlated with the observed drug release. Such a design could potentially be used to monitor prodrug activation in real-time and provide information regarding the location and the amount of active drug molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76427 Penicillin Amidase from Escherichia coli, 5-10 units/mg protein