EMAIL THIS PAGE TO A FRIEND
European review for medical and pharmacological sciences
Acute myocardial infarction during the early postpartum period successfully treated with tirofiban.
PMID 22582479
Abstract
Acute myocardial infarction (AMI) during the early postpartum period is rare but may be associated with poor maternal outcome. We report an inferior AMI in 30-year-old woman with mitral valve replacement during early postpartum period successfully treated with tirofiban. For a patient that has got AMI due to massive thrombus and that is not suitable for percutaneous coronary intervention and has got contraindication to thrombolytic therapy, glikoprotein IIb-IIIa inhibitor (tirofiban) infusion must be kept in mind.