EMAIL THIS PAGE TO A FRIEND
The Journal of urology
Injection therapy for reflux: why it works and why it fails.
PMID 22583639
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000744 Dextranomer, European Pharmacopoeia (EP) Reference Standard