EMAIL THIS PAGE TO A FRIEND
Journal of molecular modeling
DFT studies on one-electron oxidation and one-electron reduction for 2- and 4-aminopyridines.
PMID 22585356
Abstract
Quantum-chemical calculations {DFT(B3LYP)/6-311+G(d,p)} were performed for all possible tautomers (aromatic and nonaromatic) of neutral 2- and 4-aminopyridines and their oxidized and reduced forms. One-electron oxidation has no important effect on the tautomeric preference for 2-aminopyridine. The amine tautomer is favored. However, oxidation increases the stability of the imine NH tautomer, and its contribution in the tautomeric mixture cannot be neglected. In the case of 4-aminopyridine, one-electron oxidation increases the stability of both the amine and imine NH tautomers. Consequently, they possess very close energies. As major tautomers, they dictate the composition of the tautomeric mixture. The CH tautomers may be considered as very rare forms for both neutral and oxidized aminopyridines. A reverse situation takes place for the reduced forms of aminopyridines. One-electron reduction favors the C3 atom for the labile proton for both aminopyridines. This may partially explain the origin of the CH tautomers for the anionic states of nucleobases containing the exo NH(2) group.