EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Analysis of protein glycation using phenylboronate acrylamide gel electrophoresis.
PMID 22585480
Abstract
Carbohydrate modification of proteins adds complexity and diversity to the proteome. However, undesired carbohydrate modifications also occur in the form of glycation, resulting in diseases such as diabetes, Alzheimer's disease, autoimmune diseases, and cancer. The analysis of glycated proteins is challenging due to their complexity and variability. Numerous analytical techniques have been developed that require expensive specialised equipment and complex data analysis. In this chapter, we describe a simple electrophoresis-based method that enables users to detect, identify, and analyze these post-translational modifications. This new cost-effective methodology will aid the detection of unwanted glycation products in processed foods and may lead to new diagnostics and therapeutics for age-related chronic diseases and glycosylation disorders.