EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Hijacking a hydroxyethyl unit from a central metabolic ketose into a nonribosomal peptide assembly line.
PMID 22586110
Abstract
Nonribosomal peptide synthetases (NRPSs) usually catalyze the biosynthesis of peptide natural products by sequential selection, activation, and condensation of amino acid precursors. It was reported that some fatty acids, α-ketoacids, and α-hydroxyacids originating from amino acid metabolism as well as polyketide-derived units can also be used by NRPS assembly lines as an alternative to amino acids. Ecteinascidin 743 (ET-743), naphthyridinomycin (NDM), and quinocarcin (QNC) are three important antitumor natural products belonging to the tetrahydroisoquinoline family. Although ET-743 has been approved as an anticancer drug, the origin of an identical two-carbon (C(2)) fragment among these three antibiotics has not been elucidated despite much effort in the biosynthetic research in the past 30 y. Here we report that two unexpected two-component transketolases (TKases), NapB/NapD in the NDM biosynthetic pathway and QncN/QncL in QNC biosynthesis, catalyze the transfer of a glycolaldehyde unit from ketose to the lipoyl group to yield the glycolicacyl lipoic acid intermediate and then transfer the C(2) unit to an acyl carrier protein (ACP) to form glycolicacyl-S-ACP as an extender unit for NRPS. Our results demonstrate a unique NRPS extender unit directly derived from ketose phosphates through (α,β-dihydroxyethyl)-thiamin diphosphate and a lipoyl group-tethered ester intermediate catalyzed by the TKase-ACP platform in the context of NDM and QNC biosynthesis, all of which also highlights the biosynthesis of ET-743. This hybrid system and precursor are distinct from the previously described universal modes involving the NRPS machinery. They exemplify an alternate strategy in hybrid NRPS biochemistry and enrich the diversity of precursors for NRPS combinatorial biosynthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68138 Transketolase from E. coli, ≥0.1 U/mg