EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Examining the role of phosphate in glycosyl transfer reactions of Cellulomonas uda cellobiose phosphorylase using D-glucal as donor substrate.
PMID 22591555
Abstract
Cellobiose phosphorylase from Cellulomonas uda (CuCPase) is shown to utilize D-glucal as slow alternative donor substrate for stereospecific glycosyl transfer to inorganic phosphate, giving 2-deoxy-α-D-glucose 1-phosphate as the product. When performed in D(2)O, enzymatic phosphorolysis of D-glucal proceeds with incorporation of deuterium in equatorial position at C-2, implying a stereochemical course of reaction where substrate becomes protonated from below its six-membered ring through stereoselective re side attack at C-2. The proposed catalytic mechanism, which is supported by results of docking studies, involves direct protonation of D-glucal by the enzyme-bound phosphate, which then performs nucleophilic attack on the reactive C-1 of donor substrate. When offered D-glucose next to D-glucal and phosphate, CuCPase produces 2-deoxy-β-D-glucosyl-(1→4)-D-glucose and 2-deoxy-α-D-glucose 1-phosphate in a ratio governed by mass action of the two acceptor substrates present. Enzymatic synthesis of 2-deoxy-β-D-glucosyl-(1→4)-D-glucose is effectively promoted by catalytic concentrations of phosphate, suggesting that catalytic reaction proceeds through a quaternary complex of CuCPase, D-glucal, phosphate, and D-glucose. Conversion of D-glucal and phosphate presents a convenient single-step synthesis of 2-deoxy-α-D-glucose 1-phosphate that is difficult to prepare chemically.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
464058
D-Glucal, 96%
C6H10O4