EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Sugar consumption and ethanol fermentation by transporter-overexpressed xylose-metabolizing Saccharomyces cerevisiae harboring a xyloseisomerase pathway.
PMID 22591844
Abstract
Four kinds of transporters, HXT1 and HXY7 from Saccharomyces cerevisiae, and GXF1 and GXS1 from Candida intermedia, were overexpressed in xylose-metabolizing S. cerevisiae harboring a xyloseisomerase-based pathway. Overexpression of transporter enhanced sugar consumption and ethanol production, and GXF1 was efficient for ethanol fermentation from both glucose and xylose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4166 Glucose Isomerase from Streptomyces murinus, ≥350 U/g