EMAIL THIS PAGE TO A FRIEND
The Consultant pharmacist : the journal of the American Society of Consultant Pharmacists
Aspirin: a history, a love story.
PMID 22591976
Abstract
Most pharmacists know that aspirin's origins lie with willow bark, but they may be unaware of its role in the development of the pharmaceutical industry. Evolving from salacin (the active ingredient in many plant remedies) to salicylic acid (an analgesic in its own right) to the more effective, less toxic acetylsalicylic acid, this pain reliever cornered the nonsteroidal anti-inflammatory market for more than 70 years. It helped the dye industry branch into pharmaceuticals, and is now used in multiple indications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0625
D-(−)-Salicin, ≥99% (GC)
C13H18O7