EMAIL THIS PAGE TO A FRIEND
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Famotidine suppresses osteogenic differentiation of tendon cells in vitro and pathological calcification of tendon in vivo.
PMID 22592911
Abstract
Heterotopic ossification or calcification follows any type of musculoskeletal trauma and is known to occur after arthroplasties of hip, knee, shoulder, or elbow; fractures; joint dislocations; or tendon ruptures. Histamine receptor H2 (Hrh2) has been shown to be effective for reducing pain and decreasing calcification in patients with calcifying tendinitis, which suggested that H2 blockers were effective for the treatment of tendon ossification or calcification. However, the detailed mechanisms of its action on tendon remain to be clarified. We investigated the mechanisms underlying H2 blocker-mediated suppression of tendon calcification, with a focus on the direct action of the drug on tendon cells. Famotidine treatment suppressed the mRNA expressions of Col10a1 and osteocalcin, ossification markers, in a tendon-derived cell line TT-D6, as well as a preosteoblastic one MC3T3-E1. Both of the cell lines expressed Hrh2; histamine treatment induced osteocalcin expression in these cells. Famotidine administration suppressed calcification in the Achilles tendon of ttw mice, a mouse model of ectopic ossification. These data suggest that famotidine inhibits osteogenic differentiation of tendon cells in vitro, and this inhibition may underlie the anti-calcification effects of the drug in vivo. This study points to the use of H2 blockers as a promising strategy for treating heterotopic ossification or calcification in tendon, and provides evidence in support of the clinical use of famotidine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6889
Famotidine
C8H15N7O2S3
PHR1055
Famotidine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H15N7O2S3