EMAIL THIS PAGE TO A FRIEND
International immunopharmacology
Inhibitory effects of a spinasterol glycoside on lipopolysaccharide-induced production of nitric oxide and proinflammatory cytokines via down-regulating MAP kinase pathways and NF-κB activation in RAW264.7 macrophage cells.
PMID 22595195
Abstract
Extracts from the leaves of Stewartia koreana are known to exhibit strong anti-inflammatory activity. Investigation of bioactive compounds from S. koreana has led to the isolation of 3-O-β-d-glucopyanosylspinasterol (spinasterol-Glc), a spinasterol glycoside. In the present study, we examined the effects of spinasterol-Glc on production of nitric oxide (NO) and proinflammatory cytokines in LPS-treated RAW264.7 macrophage cells and in mouse models. Our results showed that spinasterol-Glc inhibited the production of NO and proinflammatory cytokines such as TNF-α, IL-6 and IL-1β in dose-dependent manners in LPS-treated RAW264.7 cells. Spinasterol-Glc inhibited the expression of iNOS and the proinflammatory cytokine genes. Spinasterol-Glc also inhibited phosphorylation of IκB-α and IKKα/β as well as translocation of NF-κB to the nucleus. We demonstrated that spinasterol-Glc reduced transcription of the NF-κB minimal promoter and NF-κB DNA binding activity. Administration of the spinasterol-Glc significantly decreased the plasma levels of these inflammatory mediators including TNF-α, IL-6 and IL-1β in LPS-injected mice and improved survival of septic mice with lethal endotoxemia. These results suggest that spinasterol-Glc has effective inhibitory effects on production of inflammatory mediators via inhibition of MAP kinases/NF-κB activities, and can be used as a potential anti-inflammatory agent for the prevention and treatment of inflammatory diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47132
Stigmasterol, certified reference material, 10 mg/mL in chloroform
C29H48O