EMAIL THIS PAGE TO A FRIEND
Connective tissue research
Effect of norcantharidin on the human breast cancer Bcap-37 cells.
PMID 22606958
Abstract
Norcantharidin (NCTD), a chemically modified form of cantharidin, is a potential anticancer drug. In this study, the effects of NCTD on the cellular viability, reactive oxygen species (ROS), mitochondrial membrane potential (MMP), and DNA damage in the human breast cancer cell line Bcap-37 were investigated with confocal and fluorescence microscopy. The cell cycle was further analyzed using the CellQuest software of a Becton-Dickinson FACS flow cytometer. The results indicated that the cellular viability was decreased with the growing concentrations of NCTD and time exposure. Moreover, the fluorescence intensity of ROS was increased, whereas the MMP was decreased in Bcap-37 cells with the growing concentrations of NCTD. NCTD induced a dose-dependent DNA damage and reduced the G1 peak in Bcap-37 cells. The G2/M peak of Bcap-37 was also decreased by the higher concentration of NCTD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8784
Norcantharidin, solid
C8H8O4