EMAIL THIS PAGE TO A FRIEND
Clinical and experimental dermatology
Pseudofolliculitis barbae induced by oral minoxidil.
PMID 22607496
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2300000
Minoxidil, European Pharmacopoeia (EP) Reference Standard
C9H15N5O