EMAIL THIS PAGE TO A FRIEND
Experimental and molecular pathology
A model for cardiomyocyte cell death: insights into mechanisms of oncosis.
PMID 22609242
Abstract
It is now known that there are at least two basic patterns of cell injury progressing to cell death: cell injury with swelling, known as oncosis, and cell injury with shrinkage, known as apoptosis. Both types of cell death are "programmed" in the sense that the genetic information and many of the enzymes and other factors pre-exist in the cell. Previous investigation has pointed to cardiomyocyte ischemic injury evolving as the oncotic pattern of injury, although apoptosis has also been implicated. This study was designed, using a unique cell model system, to gain insight into the molecular events of anticancer agent-induced cardiomyocyte injury. Cardiomyocytes exposed for 2 h to 1.5 μg/ml sanguinarine consistently displayed the morphology of apoptosis in over 80% of cells, whereas a higher dose of 25 μg/ml at 2 h yielded the pattern of oncosis in over 90% of cells. Microarray analysis revealed altered expression of 2514 probes in sanguinarine-induced oncosis and 1643 probes in apoptosis at a level of significance of p<0.001. Some of the inductions such as perforin were found to be higher than 11-fold in oncosis. When perforin was blocked by perforin-specific siRNA we found a reduction in oncotic cell death. These results strengthen the notion that oncosis is not representative of nonspecific necrosis, but constitutes a genetically controlled form of "programmed cell death"; and also that oncosis might represent a pathogenetic mechanism of cardiomyocyte injury. This is also the first demonstration of the involvement of perforin in cardiomyocyte oncosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5890
Sanguinarine chloride hydrate, ≥98% (HPLC)
C20H14ClNO4 · xH2O