EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
18β-Glycyrrhetinic acid preferentially blocks late Na current generated by ΔKPQ Nav1.5 channels.
PMID 22609834
Abstract
To compare the effects of two stereoisomeric forms of glycyrrhetinic acid on different components of Na(+) current, HERG and Kv1.5 channel currents. Wild-type (WT) and long QT syndrome type 3 (LQT-3) mutant ΔKPQ Nav1.5 channels, as well as HERG and Kv1.5 channels were expressed in Xenopus oocytes. In addition, isolated human atrial myocytes were used. Two-microelectrode voltage-clamp technique was used to record the voltage-activated currents. Superfusion of 18β-glycyrrhetinic acid (18β-GA, 1-100 μmol/L) blocked both the peak current (I(Na,P)) and late current (I(Na,L)) generated by WT and ΔKPQ Nav1.5 channels in a concentration-dependent manner, while 18α-glycyrrhetinic acid (18α-GA) at the same concentrations had no effects. 18β-GA preferentially blocked I(Na,L) (IC(50)=37.2 ± 14.4 μmol/L) to I(Na,P) (IC(50)=100.4 ± 11.2 μmol/L) generated by ΔKPQ Nav1.5 channels. In human atrial myocytes, 18β-GA (30 μmol/L) inhibited 47% of I(Na,P) and 87% of I(Na,L) induced by Anemonia sulcata toxin (ATX-II, 30 nmol/L). Superfusion of 18β-GA (100 μmol/L) had no effects on HERG and Kv1.5 channel currents. 18β-GA preferentially blocked the late Na current without affecting HERG and Kv1.5 channels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8503
18α-Glycyrrhetinic acid, ≥95%
C30H46O4