EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Expression of bacterial GshF in Pichia pastoris for glutathione production.
PMID 22610434
Abstract
Conventionally, two consecutive enzymatic reactions catalyzed by γ-glutamylcysteine synthetase and glutathione synthetase are most commonly used for glutathione production. Here we demonstrate that bacterial bifunctional GshF can be used for glutathione production in a eukaryotic system without accumulation of the intermediate γ-glutamylcysteine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0903
γ-Glu-Cys, ≥80% (HPLC)
C8H14N2O5S