EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Chromogen-based immunohistochemical method for elucidation of the coexpression of two antigens using antibodies from the same species.
PMID 22610462
Abstract
The authors established a chromogen-based, double immunolabeling method using antibodies from the same species without any unwanted cross-reactivity. In addition, time-consuming staining steps were shortened by using polymer-based secondary antibodies. Taking advantage of the nature of the chromogen 3-amino-9-ethylcarbazole (AEC), which is used as a horseradish peroxidase substrate for antibody detection, the AEC-derived signals in the first color development were easily eliminated by alcohol treatment. Therefore, the signals from the first staining did not interfere with those from the subsequent second staining, which used the chromogen 3,3'-diaminobenzidine. The co-localization of antigens within the same cell could be confirmed using this method, because cell images of the individual dye staining steps could be obtained and developed. The images from each step could be expressed in pseudo-colors in a dark field by using a computer. As a result, merged images could be constructed that resembled the images acquired by the fluorescent immunolabeling technique. The resolution of this method enabled analysis of the coexpression of two antigens in the same cell in the same section. The authors have named this staining technique the elucidation of the coexpression of two antigens in a cell using antibodies from the same species (ECSS).