EMAIL THIS PAGE TO A FRIEND
Wei sheng yan jiu = Journal of hygiene research
[Effects of citreoviridin on the expression of MCP-1 and ILs induced by TNF-alpha in vein endothelial cells].
PMID 22611924
Abstract
To investigate the effects of citreoviridin (CIT) on the expression of MCP-1, IL-1beta, IL-6 and IL-8 induced by TNF-alpha in human umbilical vein endothelial cells (HUVECs). HUVECs isolated from the umbilical cord of neonates within 1 hour after birth (informed with consent form) were cultured in DMEM/ F12 media. After 80% of HUVECs were confluent, the cells were divided into four groups and treated with CIT (2 micromol/L) and/or TNF-alpha (10 microg/L). The levels of MCP-1, IL-1beta, IL-6 and IL-8 in the supernatant of cell culture media were measured by ELISA, the activation of NF-kappaB in HUVECs was detected by immunofluorescence staining, and the expression of MCP-1 mRNA was determinated by RT-PCR assay. The levels of MCP-1, IL-1beta, IL-6 and IL-8 in the supernatant and the expression of NF-kappaB P65 and MCP-1 mRNA of HUVECs were higher in TNF-alpha group and TNF-alpha + CIT group than those in the control group (P < 0.05), and those of TNF-alpha + CIT group was higher than TNF-alpha group (P < 0.05). The expression of NF-kappaB, MCP-1, IL-1beta, IL-6 and IL-8 in HUVECs up-regulated by TNF-alpha was promoted by CIT.