EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Nickel-catalyzed carboxylation of aryl and vinyl chlorides employing carbon dioxide.
PMID 22612592
Abstract
Nickel-catalyzed carboxylation of aryl and vinyl chlorides employing carbon dioxide has been developed. The reactions proceeded under a CO(2) pressure of 1 atm at room temperature in the presence of nickel catalysts and Mn powder as a reducing agent. Various aryl chlorides could be converted to the corresponding carboxylic acid in good to high yields. Furthermore, vinyl chlorides were successfully carboxylated with CO(2). Mechanistic study suggests that Ni(I) species is involved in the catalytic cycle.