EMAIL THIS PAGE TO A FRIEND
New biotechnology
Efficacy of pentane, toluene, and benzene to support aerobic cometabolism of ethylene dibromide.
PMID 22613211
Abstract
The ability of pentane, benzene, and toluene to support aerobic cometabolism of ethylene dibromide (1,2-dibromoethane, EDB) was evaluated. A pentane enrichment culture cometabolized EDB, with a transformation capacity of 0.35 μmol EDB/mg biomass (66.2 μg EDB/mg biomass) in the absence of growth substrate. It also cometabolized EDB while actively growing on pentane. However, enrichment cultures grown on benzene or toluene could not cometabolize EDB, with or without their respective growth substrates.