EMAIL THIS PAGE TO A FRIEND
ChemMedChem
6-halogenochromones bearing tryptamine: one-step access to potent and highly selective inhibitors of breast cancer resistance protein.
PMID 22615192
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
193747
Tryptamine, ≥97%
C10H12N2
246557
Tryptamine hydrochloride, 99%
C10H12N2 · HCl