EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Biological pretreatment for production of lignocellulosic biofuel.
PMID 22617032
Abstract
Lignocellulosic biomass was submitted to a biological pretreatment prior to a catalytic hydroliquefaction in order to produce biofuel. The biodegradation process was conducted over 3 months in a reactor under controlled conditions. During the biodegradation process the organic matter was characterised and its evolution was correlated with physico-chemical parameters. In parallel with the analysis of the lipidic fraction, analytical pyrolysis was used to monitor bacterial activity. The alterations of branched to linear fatty acids ratio and of mono- to diacids ratio were compared when determined by thermochemolysis and observed in the directly extractable lipids. The evolution of the phytol to the corresponding isoprenoic ketone ratio was observed to be dependent on the desorption technique since it decreases using headspace while it increases using pyrolysis. "Humic"/"fulvic acids" ratio, infrared spectroscopy and thermodifferential analysis were used to determine the degree of OM complexification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87708
Tetramethylammonium bromide, for ion pair chromatography, ≥99.0% (AT)
C4H12BrN
426296
Tetramethylammonium bromide, ACS reagent, ≥98.0%
C4H12BrN
195758
Tetramethylammonium bromide, 98%
C4H12BrN
87709
Tetramethylammonium bromide, for electrochemical analysis, ≥99.0%
C4H12BrN
87718
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
C4H12ClN
74202
Tetramethylammonium chloride, for ion pair chromatography, ≥99.0% (AT)
C4H12ClN
T19526
Tetramethylammonium chloride, reagent grade, ≥98%
C4H12ClN
T3411
Tetramethylammonium chloride solution, for molecular biology
459135
Tetramethylammonium fluoride, 97%
C4H12FN
T7505
Tetramethylammonium hydroxide pentahydrate, ≥97%
C4H13NO · 5H2O
87741
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
C4H13NO · 5H2O
426318
Tetramethylammonium hydroxide solution, ACS reagent
C4H13NO
334901
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
C4H13NO
331635
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
C4H13NO
328251
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
C4H13NO
235946
Tetramethylammonium iodide, 99%
C4H12IN
425257
Tetramethylammonium nitrate, 95%
C4H12N2O3
02799
Tetramethylammonium sulfate, for ion pair chromatography, ≥99.0% (T)
C8H24N2O4S
87743
Tetramethylammonium sulfate hydrate
C8H24N2O4S · xH2O