EMAIL THIS PAGE TO A FRIEND
The journal of ECT
Treatment of post-electroconvulsive therapy headache with topical methyl salicylate.
PMID 22622298
Abstract
Headache after administration of electroconvulsive therapy (ECT) is common, affecting approximately half of patients treated. Post-ECT headache is typically treated with acetaminophen or nonsteroidal anti-inflammatory drugs but occasionally requires agents such as sumatriptan, opioids, or β-blockers. We report on a patient whose severe post-ECT headaches responded completely to methyl salicylate ointment, applied to the area of his temporalis and masseter muscles. Topical methyl salicylate is generally well tolerated and may be a viable option for some patients with post-ECT headache.