EMAIL THIS PAGE TO A FRIEND
Proteomics
Roles of interferon-gamma and its target genes in schizophrenia: Proteomics-based reverse genetics from mouse to human.
PMID 22623148
Abstract
A decreased production of interferon gamma (IFNG) has been observed in acute schizophrenia. In order to explore the possible relationship between IFNG and schizophrenia, we attempted to analyze the differentially expressed proteins in the brains of interferon-gamma knockout (Ifng-KO) mice. Five upregulated and five downregulated proteins were identified with 2D gels and MALDI-TOF/TOF MS analyses in Ifng-KO mouse brain. Of the identified proteins, we focused on creatine kinase brain (CKB) and triose phosphate isomerase 1 (TPI1). Consistent with the proteomic data, reverse transcriptase-mediated PCR, immunoblotting, and immunohistochemistry analyses confirmed that the levels of gene expressions of Ckb and Tpi1 were downregulated and upregulated, respectively. When we analyzed the genetic polymorphisms of the single nucleotide polymorphisms (SNPs) of their human orthologous genes in a Korean population, the promoter SNPs of CKB and TPI1 were weakly associated with schizophrenia. In addition, IFNG polymorphisms were associated with schizophrenia. These results suggest that IFNG and proteins affected by IFNG may play a role in the pathogenesis of schizophrenia.