EMAIL THIS PAGE TO A FRIEND
Journal of food science
Effect of moisture, lipids, and select amino acid blocking agents on the formation and stability of metastable radicals in powdered soy proteins.
PMID 22625876
Abstract
Incremental increases in the moisture content of powdered soy protein products from 4.4% to 13.4% produced an inverse effect on the ability of soy proteins to maintain metastable free radicals. The corresponding reduction in electron paramagnetic resonance signal was not due to dielectric loss in the range of moisture contents examined. Subsequent evaluations of various treatments were conducted after drying soy proteins with molecular sieve to a water activity below 0.085 in order to minimize the influence from variations in moisture. Isolated soy protein (ISP) samples, prepared with "defatted flour" that had been further extracted with chloroform/methanol (2: 1), had a 96% reduction in total lipids compared to the control ISP samples. The initial rate of radical accumulation in the "reduced-lipid" ISP for the first 3 wk was not significantly different from the initial rate of radical increases in the control ISP. After 3 wk, radical accumulation in the "reduced-lipid" ISP continued to increase, but at a rate that was less than the control. These findings indicate that the initial reactions contributing to the formation of metastable radicals in the powdered ISP are not strongly dependent on associated lipids. Blocking sulfhydryl groups during ISP preparation with N-ethylmaleimide did not significantly slow the rate of radical accumulation compared to the control ISP. Blocking arginine residues in ISP samples with phenylglyoxal caused an increased rate of radical accumulation for the first 4 wk. Levels of metastable radicals in powdered soy protein products typically range from 10 to 100 times greater than the free radicals in other food protein sources. This current research examines various compositional and treatment parameters that might be used to minimize the content of free radicals in foods containing soy proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78600
Phenylglyoxal monohydrate, crystallized, ≥97.0% (GC)
C8H6O2 · H2O