EMAIL THIS PAGE TO A FRIEND
Biomeditsinskaia khimiia
[Phenothiazines are slowly oxidizable substrates of horseradish peroxidase].
PMID 22629605
Abstract
Reactions of peroxidase oxidation of triftazine and thioproperazine have been investigated in the presence of horseradish peroxidase using steady state kinetic methods. It has been shown that phenothiazines are slowly oxidizable substrates for horseradish peroxidase. k(cat) and K(m) values have been determined in the range of pH from 4.5 to 7.5. The study of co-oxidation of phenothiazines and o-dianisidine (ODN) revealed that in the presence of aminazine and ODN in the reaction medium both substances follow sequential oxidation. ODN oxidation was not observed until full conversion of aminazine. At pH 4.5-5.5 thioproperazine bound to the enzyme-substrate complex and caused a nticompetitive inhibition of peroxidase. At pH>5.5 sequential substrate oxidation with preferential thioproperazine conversion occurred. In the range of pH from 4.5 to 7.5 triftazine did not influence ODN oxidation.
Related Materials