EMAIL THIS PAGE TO A FRIEND
Mikrobiologiia
[Biodegradation of glyphosate by soil bacteria: optimization of culture and method for active biomass preservation].
PMID 22629681
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
289868
Methylphosphonic acid, 98%
CH5O3P