EMAIL THIS PAGE TO A FRIEND
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP
Activity of beta-lactam beta-lactamase inhibitor combinations against extended spectrum Beta-lactamase producing Enterobacteriaceae in urinary isolates.
PMID 22630093
Abstract
To determine the susceptibility pattern of beta-lactam beta-lactamase inhibitor combinations against extended-spectrum beta-lactamase (ESBL) producing Enterobacteriaceae in urinary isolates. Observational study. Ziauddin University Hospital, Karachi, from February to October 2008. A total of 190 consecutive non-duplicate isolates of ESBL producing Enterobacteriaceae from urine samples of in-patients were included in the study. Urinary samples from out-patients, repeat samples and non-ESBL producing isolates were excluded. Detection of ESBL was carried out by double disk diffusion technique. Antimicrobial susceptibility testing was performed using modified Kirby Bauer's disk diffusion method according to CLSI guidelines. Statistical analysis was performed by SPSS version 10. Of the 190 ESBL isolates tested, 88 cases (46.31%) were sensitive and 6 cases (3.15%) were resistant to all three combinations, the rest 96 cases (50.52%) were resistant to at least one of the combinations. Susceptibility pattern of cefoperazone/sulbactam, piperacillin/tazobactam, and amoxicillin/clavulanic acid was 95.26, 92.10, and 44.31 percent respectively. Cefoperazone/sulbactam exhibited the best activity against ESBL producing Enterobacteriaceae followed by piperacillin/tazobactam. Hospital antibiotic policies should be reviewed periodically to reduce the usage of extended spectrum cephalosporins and replace them with beta-lactam beta-lactamase inhibitor combinations agent for treating urinary tract infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4292
Cefoperazone sodium salt, 870-1015 μg/mg (anhydrous basis)
C25H26N9NaO8S2