EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Novel method to reduce fishy aftertaste in wine and seafood pairing using alcohol-treated yeast cells.
PMID 22630330
Abstract
"Fishy aftertaste" is sometimes perceived in wine consumed with seafood. Iron in wine has been reported to be a key compound that produces fishy aftertaste. However, cost-effective methods to remove iron from wine have not been developed. Here, we describe a cost-effective and safe iron adsorbent consisting of alcohol-treated yeast (ATY) cells based on the observation that nonviable cells adsorbed iron after completion of fermentation. Treatment of cells with more than 40% (v/v) ethanol killed them without compromising their ability to adsorb iron. Drying the ATY cells did not reduce iron adsorption. Use of ATY cells together with phytic acid had a synergistic effect on iron removal. We term this means of removing iron the "ATY-PA" method. Sensory analysis indicated that fishy aftertaste in wine-seafood pairings was not perceived if the wine had been pretreated with both ATY cells and phytic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5681
Phytic acid dipotassium salt
C6H16K2O24P6