EMAIL THIS PAGE TO A FRIEND
Infectious disease clinics of North America
American trypanosomiasis (Chagas disease).
PMID 22632639
Abstract
Chagas disease, also known as American trypanosomiasis, is a chronic infection caused by Trypanosoma cruzi, a protozoan parasite. It is transmitted to human beings mainly through the feces of infected triatomine bugs. The disease affects an estimated 8 to 10 million people in the Americas, putting them at risk of developing life-threatening cardiac and gastrointestinal complications. This article provides a brief update on the epidemiology, clinical manifestations, diagnosis, and treatment of Chagas disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3415
Nifurtimox, ≥98% (HPLC)
C10H13N3O5S