EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Reactive oxygen species mediated ginsenoside Rg3- and Rh2-induced apoptosis in hepatoma cells through mitochondrial signaling pathways.
PMID 22634290
Abstract
Panax ginseng (P. ginseng) has anti-cancer effects in several cancer models. Ginsenosides are the main bioactive components in P. ginseng. Korean red ginseng (KRG) extract can potently kill various cancer cells and ginsenosides Rg3 (GRg3) and Rh2 (GRh2) are the primary ginsenosides in KRG. This study was carried out to examine whether KRG and its primary ginsenosides (GRg3 and GRh2) affect apoptosis of human hepatocellular carcinoma cells (Hep3B). KRG, GRg3 and GRh2 have obvious cytotoxic and apoptotic effects in Hep3B cells as evidenced by a decrease in cell viability and mitochondria membrane potential, but an increase in LDH release. In the mitochondria-mediated apoptosis pathway, KRG, GRg3 and GRh2 have the ability to stimulate the release of mitochondrial cytochrome c, activation of caspase-3 and Bax protein, inhibition of Bcl-2 protein and production of intracellular reactive oxygen species in Hep3B cells. These results suggest that KRG, GRg3 and GRh2 may induce apoptosis by direct activation of the mitochondrial pathway.
Related Materials