EMAIL THIS PAGE TO A FRIEND
Medicinski glasnik : official publication of the Medical Association of Zenica-Doboj Canton, Bosnia and Herzegovina
Starch for health.
PMID 22634903
Abstract
It is well established that part of starch is resistant to human amylases and escapes undigested to large bowel. This fraction of starch is resistant starch. Recent studies have shown that resistant starch may be a substrate for bacterial flora of the colon and serves as prebiotic. Short chain fatty acids (SCFA) produced by colonic fermentation of resistant starch may have impact on colonic function and health of humans. This paper summarises current knowledge on properties and health impact of resistant starch.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2388
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10