EMAIL THIS PAGE TO A FRIEND
Methods in Molecular Biology
Dissociation of embryonic kidney followed by re-aggregation as a method for chimeric analysis.
PMID 22639257
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB2219
Anti-Aquaporin 1 Antibody, clone, 1.0 mg/mL, Chemicon®