EMAIL THIS PAGE TO A FRIEND
Klinichna khirurhiia
[Comparative estimation of the application efficacy for ceftriaxon and Hepacef combi preparations in the abdominal infection treatment].
PMID 22642085
Abstract
A positive experience was described in the work, concerning application of Hepacef combi (cefoperazon/sulbactam, manufactured by "ARTERIUM" (Corporation, Ukraine) in the treatment of inflammatory-purulent diseases of abdominal organs as a method of empirical therapy. There was the preparation action efficacy estimated together with the diseases outcomes analysis, and possible recommendations for its application suggested. The preparation may be recommended for application as the empirical therapy agent in the abdominal infection treatment.