EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Effect of rifampin and rifabutin on the pharmacokinetics of lersivirine and effect of lersivirine on the pharmacokinetics of rifabutin and 25-O-desacetyl-rifabutin in healthy subjects.
PMID 22644026
Abstract
Lersivirine is a nonnucleoside reverse transcriptase inhibitor (NNRTI) with a unique resistance profile exhibiting potent antiviral activity against wild-type HIV and several clinically relevant NNRTI-resistant strains. Lersivirine, a weak inducer of the cytochrome P450 (CYP) enzyme CYP3A4, is metabolized by CYP3A4 and UDP glucuronosyltransferase 2B7 (UGT2B7). Two open, randomized, two-way (study 1; study A5271008) or three-way (study 2; study A5271043) crossover phase I studies were carried out under steady-state conditions in healthy subjects. Study 1 (n = 17) investigated the effect of oral rifampin on the pharmacokinetics (PKs) of lersivirine. Study 2 (n = 18) investigated the effect of oral rifabutin on the PKs of lersivirine and the effect of lersivirine on the PKs of rifabutin and its active metabolite, 25-O-desacetyl-rifabutin. Coadministration with rifampin decreased the profile of the lersivirine area under the plasma concentration-time curve from time zero to 24 h postdose (AUC(24)), maximum plasma concentration (C(max)), and plasma concentration observed at 24 h postdose (C(24)) by 85% (90% confidence interval [CI], 83, 87), 83% (90% CI, 79, 85), and 92% (90% CI, 89, 94), respectively, versus the values for lersivirine alone. Coadministration with rifabutin decreased the lersivirine AUC(24), C(max), and C(24) by 34% (90% CI, 29, 39), 25% (90% CI, 16, 33), and 58% (90% CI, 52, 64), respectively, compared with the values for lersivirine alone. Neither the rifabutin concentration profile nor overall exposure was affected following coadministration with lersivirine. Lersivirine and rifabutin reduced the 25-O-desacetyl-rifabutin AUC(24) by 27% (90% CI, 21, 32) and C(max) by 27% (90% CI, 19, 34). Lersivirine should not be coadministered with rifampin, which is a potent inducer of CYP3A4, UGT2B7, and P-glycoprotein activity and thus substantially lowers lersivirine exposure. No dose adjustment of rifabutin is necessary in the presence of lersivirine; an upward dose adjustment of lersivirine may be warranted when it is coadministered with rifabutin.