EMAIL THIS PAGE TO A FRIEND
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology
Polytherapy for migraine prophylaxis.
PMID 22644191
Abstract
Migraine is a chronic neurological disorder with episodic manifestations, progressive in some individuals. Preventive treatment is recommended for patients with frequent or disabling attacks. A sizeable proportion of migraineurs in need of preventive treatment does not significantly benefit from monotherapy. This short review evaluates the role of pharmacological polytherapy in migraine prevention.