EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Competitive inhibition of cytosolic Ca2+-dependent phospholipase A2 by acteoside in RBL-2H3 cells.
PMID 22644858
Abstract
The aim of this study was to investigate whether acteoside isolated from Clerodendron trichotomum Thunberg may act as a selective inhibitor of phospholipase A(2) in RBL-2H3 cells. Acteoside dose-dependently inhibited 0.5 μM melittin-induced release of [(3)H]arachidonic acid, which was due to the inhibition of cytosolic Ca(2+)-dependent phospholipase A(2) (cPLA(2)) rather than secretory PLA(2) (sPLA(2)). In Dixon plots, the apparent K ( i ) value of acteoside on cPLA(2) was 5.9 μM and the inhibitory pattern appeared to be a competitive inhibitor. The above data, suggests that acteoside acts as a competitive inhibitor of cPLA(2) in RBL-2H3 cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V4015
Verbascoside, ≥99% (HPLC)
C29H36O15
00820580
Verbascoside, primary reference standard
C29H36O15