EMAIL THIS PAGE TO A FRIEND
European journal of clinical pharmacology
Interaction between amorolfine and acenocoumarol.
PMID 22648278
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0074
Acenocoumarol, ≥98% (HPLC)
C19H15NO6