EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Alperujo extract, hydroxytyrosol, and 3,4-dihydroxyphenylglycol are bioavailable and have antioxidant properties in vitamin E-deficient rats--a proteomics and network analysis approach.
PMID 22648667
Abstract
Olive products are rich in phenolic compounds, which are natural antioxidants in vitro. We tested the in vivo effects of alperujo, an olive production by-product, as well as hydroxytyrosol and 3,4-dihydroxyphenylglycol (DHPG) isolated from alperujo, on indices and pathways of oxidative and metabolic stress in a vitamin E-deficient rat model. Rats were fed a vitamin E-deficient diet for 10 weeks, followed by this diet supplemented with either 100 mg/kg diet dα-tocopherol, alperujo extract, hydroxytyrosol, or 10 mg/kg diet DHPG, for a further 2 weeks. We detected alperujo phenolics in tissues and blood, indicating they are bioavailable. Alperujo extract partially ameliorated elevated plasma levels of thiobarbituric acid reactive substances and also lowered plasma cholesterol levels, whereas hydroxytyrosol increased plasma triglyceride levels. Proteomics and subsequent network analysis revealed that hepatic mitochondrial aldehyde dehydrogenase (ALDH2), of which protein and activity levels were regulated by dα-tocopherol and olive phenolics, represents a novel central regulatory protein hub affected by the dietary interventions. The in vivo free radical scavenging properties of olive phenolics appear relatively modest in our model. But alternative mechanisms, including regulation of ALDH2, may represent relevant antioxidant mechanisms by which dietary olive phenolics could have beneficial impact on cardiovascular health.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4291
3-Hydroxytyrosol, ≥98% (HPLC)
C8H10O3
D9753
DL-3,4-Dihydroxyphenyl glycol
C8H10O4