EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Giant-cell tumor of the tendon sheath in the upper cervical spine.
PMID 22649149