EMAIL THIS PAGE TO A FRIEND
TheScientificWorldJournal
Colorectal cancer chemoprevention: is this the future of colorectal cancer prevention?
PMID 22649288
Abstract
Colorectal cancer (CRC) is presently one of the most common causes of cancer-related death in our setting and affects a great number of people each year. Screening strategies are commonly used but they do not seem enough to avoid CRC development or prevent completely its mortality. Because of this fact other prevention strategies have gained interest in recent years. Chemoprevention seems to be an attractive option in this setting and several drugs have been studied in this field. This review is focused on salicylates, nonsteroidal anti-inflammatory drugs (NSAIDs) and cycloxygenase-2 inhibitors (COXIBs), whose mechanism of action could be directly related to colon cancer chemoprevention.