EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
Allergic contact dermatitis from carmine in cosmetic blush.
PMID 22653009
Abstract
Although there are many reported cases of immediate allergy after ingestion of foods containing cochineal, there are few reports of allergic contact dermatitis from carmine. We present a rare case of allergic contact dermatitis due to carmine. A 52-year-old female presented with an itchy erythema on her cheeks at the site where blush had been applied. Patch-tested with her cosmetics, she showed a positive reaction to the blush (30% in petrolatum) and to 0.2% (but not 0.1%) carmine in petrolatum. In this case, the optimum patch-test concentration of carmine was 0.2% in petrolatum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229253
Carminic acid
C22H20O13
11298
Carminic acid, analytical standard
C22H20O13