EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
Unusual localization of allergic contact dermatitis from p-phenylenediamine.
PMID 22653013
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78460
1,4-Phenylenediamine dihydrochloride, ≥99.0%
C6H8N2 · 2HCl