EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
Butylhydroxytoluene--from jet fuels to cosmetics?
PMID 22653127
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329460 Kerosene, reagent grade, low odor
60710 Kerosene, purum